Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FPGT Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13211R
BOSSBS-13211REA 725 EUR InStock
BOSSBS-13211R
Anti-FPGT Rabbit Polyclonal Antibody
Antikörper
Guanylyltransferase enzymes transfer one molecule of GTP to another molecule and also function in the transfer of guanosine nucleotides to sugar molecules. The carbohydrate moieties that are generated are covalently attached to cell surfaces and are necessary to ensure a surface contour that satisfies a variety of physiological roles. L-fucose is an important sugar in complex carbohydrates that is frequently found on plant and mammalian N-linked glycans. FPGT (Fucose-1-phosphate guanylyltransferase), also known as GFPP (GDP-L-fucose pyrophosphorylase), is a 594 amino acid cytoplasmic protein that catalyzes the formation of GDP-L-fucose from L-fucose-1-phosphate and GTP. FPGT functions to reutilize the L-fucose that is produced uopn glycoprotein and glycolipid turnover.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: FPGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge