Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FSD2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13222R-CY3
BOSSBS-13222R-CY3EA 900 EUR InStock
BOSSBS-13222R-CY3
Anti-FSD2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
FSD2 is a 749 amino acid protein containing one B30.2/SPRY domain and two fibronectin type-III domains. The gene encoding FSD2 maps to human chromosome 15q25.2. Encoding more than 700 genes, chromosome 15 is made up of approximately 106 million base pairs and consists of about 3% of the human genome. Angelman and Prader-Willi syndromes are associated with loss of function or deletion of genes in the 15q11-q13 region. In the case of Angelman syndrome, this loss is due to inactivity of the maternal 15q11-q13 encoded UBE3A gene in the brain by either chromosomal deletion or mutation. Prader-Willi syndrome, Tay-Sachs disease and Marfan syndrome are also associated with chromosome 15.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FSD2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge