Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-AKTIP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13223R-HRP
BOSSBS-13223R-HRPEA 900 EUR InStock
BOSSBS-13223R-HRP
Anti-AKTIP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Fused toes protein homolog (FTS), also known as AKT-interacting protein (AKTIP) and Ft1, is a 292 amino acid protein that localizes to the cytoplasm and the cell membrane. A member of the ubiquitin-conjugating enzyme family, FTS binds directly to AKT1 to regulate apoptosis in a cell population. AKT1 is a protein that plays a critical role in a number of cellular responses, such as cell growth, protein synthesis, and antiapoptotic signaling. The interaction of FTS and AKT1 enhances the phosphorylation and activation of AKT1, which, through an AKT1/GSK-3∫/NFATc1 signaling cascade, results in the increased production of the proapoptotic hormone Fas ligand and thus an increase in apoptosis.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: AKTIP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge