Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FXYD6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13234R-A647
BOSSBS-13234R-A647EA 893 EUR InStock
BOSSBS-13234R-A647
Anti-FXYD6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
The mammalian FXYD family maintains Na+ and K+ gradients between the intracellular and extracellular milieus of cells in processes such as renal Na+-reabsorption, muscle contraction and neuronal excitability. FXYDs are single-span membrane proteins that share a 35 amino acid signature domain, beginning with the sequence PFXYD and containing seven invariant and six conserved amino acids. Members of the FXYD family include FXYD1 (PLM, phospholemman), FXYD2 (the g subunit of the Na+/K+-ATPase), FXYD3 (Mat8, mammary tumor protein), FXYD4 (CHIF) and FXYD5 (RIC). FXYD6 is expressed in various epithelial cells bordering the endolymph space and in the auditory neurons. FXYD6 co-localizes with Na+/K+-ATPase in the stria vascularis and can be co-immunoprecipitated with Na+/K+-ATPase. After expression, FXYD6 associates with Na+/K+-ATPase alpha1-beta1 and alpha1-beta2 isozymes, which are preferentially expressed in different regions of the inner ear and also with gastric and non-gastric H+/K+-ATPase.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: FXYD6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge