Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-FYB/ADAP Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13236R-A680
BOSSBS-13236R-A680EA 890 EUR InStock
BOSSBS-13236R-A680
Anti-FYB/ADAP Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Fyb (Fyn binding protein) and the anchoring proteins SKAP55 (src kinase-associated phosphoprotein) and SKAP55-R (SKAP55-related protein) associate with the tyrosine kinase p59fyn (13). SKAP55 and SKAP55-R bind to Fyb through their SH3 domains and function as substrates for p59Fyn in resting T cells (13). SKAP55 contains an amino-terminal pleckstrin homology domain and a carboxy-terminal SH3 domain binding motif of adjacent arginine and lysine residues followed by tandem tyrosines (i.e. RKxxYxxY) (4,5). SKAP55-R, similar in overall structure to SKAP55, contains a coiled-coil N-terminal domain (1,2). SKAP55 associates with SLAP-130, another component of the Fyn complex, which plays a role in the regulation of signaling events initiated by lymphocyte antigen receptors leading up to T cell activation (6). The human Fyb gene maps to chromosome 5p13.1 and encodes a 783 amino acid protein (7).

Type: Primary
Antigen: FYB/ADAP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 501-600/783
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge