Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-G protein alpha 13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13247R-A680
BOSSBS-13247R-A680EA 890 EUR InStock
BOSSBS-13247R-A680
Anti-G protein alpha 13 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Heterotrimeric G proteins function to relay information from cell surface receptors to intracellular effectors. Each of a very broad range of receptors specifically detects an extracellular stimulus (a photon, pheromone, odorant, hormone or neurotransmitter) while the effectors (i.e., adenyl cyclase), which act to generate one or more intracellular messengers, are less numerous. In mammals, G protein Alpha, Beta and Gamma polypeptides are encoded by at least 16, 4 and 7 genes, respectively. Most interest in G proteins has been focused on their Alpha subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Four distinct classes of G Alpha subunits have been identified; these include G Alpha s, G Alpha i, G Alpha q and G Alpha 12/13. The two members of the fourth class of G Alpha subunit proteins, G Alpha 12 and G Alpha 13, are insensitive to ADP-ribosylation by pertussis toxin, share 67% identity with each other and less than 45% identity with other G Alpha subunits and are widely expressed in a broad range of tissues.

Type: Primary
Antigen: G protein alpha 13
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 21-120/377
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge