Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GNA15 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13248R-CY5
BOSSBS-13248R-CY5EA 900 EUR InStock
BOSSBS-13248R-CY5
Anti-GNA15 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Heterotrimeric G proteins function to relay information from cell surface receptors to intracellular effectors (1). Each of a very broad range of receptors specifically detects an extracellular stimulus (a photon, pheromone, odorant, hormone or neurotransmitter) while the effectors (i.e., adenylyl cyclase), which act to generate one or more intracellular messengers, are less numerous. In mammals, G protein alpha, Beta and Gamma polypeptides are encoded by at least 16, 4 and 7 genes, respectively (2-5). Most interest in G proteins has been focused on their a subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Four distinct classes of G alpha subunits have been identified; these include Gs, Gi, Gq and Ga 12/13 (3,4). The Gi class comprises all the known a subunits that are susceptible to pertussis toxin modifications, including Ga i-1, Ga i-2, Ga i-3, Ga o, Ga t1, Ga t2, Ga z and Ga gust (4). Of these, the three Ga i subtypes function to open atrial potassium channels (6). Ga 16 is a member of the Gq subfamily and is expressed specifically in hematopoietic cells (7).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G protein alpha 16
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge