Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-G6PC3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13253R-CY5
BOSSBS-13253R-CY5EA 900 EUR InStock
BOSSBS-13253R-CY5
Anti-G6PC3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Glucose-6-phosphatase (G6Pase), is a multicomponent enzyme system that hydrolyzes glucose-6-phosphate (G6P) in the final step of gluconeogenesis and gluconeolysis. G6Pase localizes to the endoplasmic reticulum, and while liver, kidney, and intestine are the only tissues that express the first identified isoform, G6Pase-Alpha, a second form, designated G6Pase-Beta, contributes to blood glucose homeostasis in a wider range of tissues. G6Pase-Beta, also known as SCN4, UGRP or G6PC3 (glucose 6 phosphatase, catalytic, 3), is a 346 amino acid endoplasmic reticulum multi-pass membrane protein that is involved in carbohydrate biosynthesis and the gluconeogenesis pathway. Inhibited by vanadate, G6Pase-Beta hydrolyzes GP6 to glucose in the endoplasmic reticulum. Due to its necessary involvement in normal glucose metabolism, G6Pase-Beta may play an integral role in diabetes and glycogen storage diseases (GSDs).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G6PC3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge