Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GALE Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13265R-CY7
BOSSBS-13265R-CY7EA 900 EUR InStock
BOSSBS-13265R-CY7
Anti-GALE Rabbit Polyclonal Antibody (Cy7®)
Antikörper
GALE is a 348 amino acid protein that functions as the third enzyme in the Leloir pathway of galactose metabolism. A member of the sugar epimerase family, GALE exists as a homodimer, binds FAD as a cofactor and catalyzes the epimerization of UDP-N-acetylglucosamine to UDP-N-acetylgalactosamine and UDP-glucose to UDP-galactose. The gene encoding GALE maps to human chromosome 1p36.11 and mutations in this gene lead to the development of complex disorder known as epimerase-deficiency galactosemia (EDG) or galactosemia type 3, which is characterized by mental retardation, liver damage, cataracts and deafness.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GALE/Galactowaldenase
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge