Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GALK2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13267R-A488
BOSSBS-13267R-A488EA 893 EUR InStock
BOSSBS-13267R-A488
Anti-GALK2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
GALK2 is important in the first step of the galactose metabolism pathway. GALK1, which belongs to the GHMP kinase family of proteins, is a crucial enzyme for galactose metabolism, specifically converting ?d-galactose to galactose 1-phosphate. Defects in the gene encoding GALK1 can cause galactosemia II, an autosomal recessive disorder characterized by congenital cataracts during infancy, often within the first two weeks of life. In the adult population, galactosemia II can cause presenile cataracts that are secondary to accumulation of galactitol in the lens of the eye. A second gene, GALK2, encodes an enzyme with greater activity against GalNAc than galactose. GALK2 has been implicated in the salvage pathway for the reutilization of free GalNAc derived from the degradation of complex carbohydrates.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GALK2/Galactokinase 2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge