Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GALNT1/GalNAc-T1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13273R-A680
BOSSBS-13273R-A680EA 890 EUR InStock
BOSSBS-13273R-A680
Anti-GALNT1/GalNAc-T1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosamine) to serine and threonine residues onto various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T1, also known as GALNT1 (Polypeptide N-acetylgalactosaminyltransferase 1), is a ubiquitously expressed 559 amino acid single-pass type II membrane protein that localizes to the Golgi apparatus and, like other GalNAc-Ts, contains a stem region and a C-terminal ricin/lectin-like domain. GalNAc-T1 catalyzes the first reaction in O-linked oligosaccharide biosynthesis, namely the transfer of an N-acetyl-D-galactosamine residue to a protein acceptor. GalNAc-T1 uses calcium and manganese as cofactors. Due to alternative splicing events, two GalNAc-T1 isoforms are expressed.

Type: Primary
Antigen: GALNT1/GalNAc-T1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/559
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge