Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GALNT13 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13274R-A350
BOSSBS-13274R-A350EA 893 EUR InStock
BOSSBS-13274R-A350
Anti-GALNT13 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes are substrate-specific proteins that catalyze the transfer of GalNAc (N-acetylgalactosamine) to serine and threonine residues onto various proteins, thereby initiating mucin-type O-linked glycosylation in the Golgi apparatus. GalNAc-T13 (Polypeptide N-acetylgalactosaminyltransferase 13), also known as UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13, is a 556 amino acid protein that displays much stronger enzymatic activity than GalNAc-1 towards GalNAc transfer to mucin peptides such as Muc5a and Muc7. The N-terminal domain is involved in substrate binding and manganese coordination, while the C-terminal domain is involved in UDP-Gal binding and catalytic reaction. With specific expression in the central nervous system, GalNAc-T13 may be responsible for the synthesis of Tn antigen in neuronal cells, which is a universal carcinoma marker on malignant cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GALNT13/GalNAc-T13
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge