Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TUBGCP3 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13277R-CY5
BOSSBS-13277R-CY5EA 900 EUR InStock
BOSSBS-13277R-CY5
Anti-TUBGCP3 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The Gamma-Tubulin complex is composed of Gamma Tubulin and the Gamma-Tubulin complex-associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. Gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the Gamma-Tubulin complex. GCP3 (Gamma-Tubulin complex component 3), also known as TUBGCP3 or SPBC98, localizes to the centrosome and is a ubiquitously expressed 907 amino acid member of the Gamma-Tubulin complex. Like GCP2 and Gamma Tubulin, GCP3 is conserved in all eukaryotes, suggesting that it is part of a core unit involved in eukaryotic microtubule nucleation. Three isoforms of GCP3 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TUBGCP3
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge