Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-TN-C Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1327R-FITC
BOSSBS-1327R-FITCEA 883 EUR InStock
BOSSBS-1327R-FITC
Anti-TN-C Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Tenascin, also known as hexabrachion and cytotactin, is an extracellular matrix protein with a spatially and temporally restricted tissue distribution. It is a hexameric, multidomain protein with disulfide linked subunits of 190 to 240 kD, originally characterized as 'myotendinous antigen.' In the embryo it is present in dense mesenchyme surrounding developing epithelia and in developing cartilage and bone. In the adult, tenascin remains present in tendons and myotendinous junctions in the perichondrium and periosteum, as well as in smooth muscle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Tenascin C
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge