Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GAPVD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13283R-A750
BOSSBS-13283R-A750EA 890 EUR InStock
BOSSBS-13283R-A750
Anti-GAPVD1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
GAPVD1 is a 1478 amino acid peripheral membrane protein that acts both as a GTPase-activating protein (GAP) and a guanine nucleotide exchange factor (GEF). GAPVD1 participates in many processes such as insulin receptor internalization, Glut4 trafficking and endocytosis. In addition, depletion of GAPVD1 leads to delayed EGFR degradation by mediating receptor ubiquination through its RGD domain, suggesting that it may be an important mediator of carcinogenesis resulting from Ras protein mutations. There are six isoforms of GAPVD1 that are produced as a result of alternative splicing events.

Type: Primary
Antigen: GAPVD1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1151-1250/1478
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge