Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GBA3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13298R-CY7
BOSSBS-13298R-CY7EA 900 EUR InStock
BOSSBS-13298R-CY7
Anti-GBA3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
CBG is a monomeric enzyme involved in the absorption and metabolism of flavonoid glucosides. CBG is found predominately in the liver, but is also located in tissues such as spleen, small intestine and kidney. Through its catalytic activity, CBG is able to hydrolyze a variety of glycosides including phytoestrogens, cyanogens, and flavonols. Although its catalytic activity extends to many dietary flavonoids, CBG has increased specificity for hydrophobic aglycones such as beta-D-glucoside and beta-D-galactoside. Hydrolysis is inhibited by sodium taurocholate and glucosyl-sphingosine, both of which regulate CBG enzymatic activity. Deficiencies in CBG have been implicated in Gaucher’s disease, a lysosomal storage disease that causes a build up of fatty material in the spleen, liver, lung and kidneys.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GBA3/CBG
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge