Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GBAS Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13299R-CY7
BOSSBS-13299R-CY7EA 900 EUR InStock
BOSSBS-13299R-CY7
Anti-GBAS Rabbit Polyclonal Antibody (Cy7®)
Antikörper
NIPSNAP2 is a 286 amino acid protein that is abundantly expressed in heart and skeletal muscle. Belonging to the NIPSNAP family, NIPSNAP2 may be involved in vesicular transport. NIPSNAP2 contains a signal peptide, a transmembrane domain and two tyrosine phosphorylation sites. NIPSNAP2 is encoded by a gene mapping to human chromosome 7p11.2. Chromosomal region 7p12 is amplified in approximately 40% of glioblastomas, the most common and malignant form of central nervous system tumor. Human chromosome 7 houses over 1,000 genes and comprises nearly 5% of the human genome. Defects in some of the genes localized to chromosome 7 have been linked to Osteogenesis imperfecta, Williams-Beuren syndrome, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GBAS
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge