Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GBE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13300R-HRP
BOSSBS-13300R-HRPEA 900 EUR InStock
BOSSBS-13300R-HRP
Anti-GBE1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
GBE1 is a 702 amino acid protein that is expressed at high levels in muscle and liver and is involved in glycogen biosynthesis. Existing as a monomer, GBE1 catalyzes the transfer of alpha-1,4-linked glucosyl units from the outer end of a glycogen chain to an alpha-1,6 position on a neighboring glycogen chain and, via this catalytic activity, plays an essential role in glycogen accumulation. Defects in the gene encoding GBE1 are the cause of glycogen storage disease type 4 (GSD4) and adult polyglucosan body disease (APBD), the first of which is a metabolic disorder that is associated with the accumulation of polysaccharides and is characterized by liver disease during childhood. Unlike GSD4, APBD is a late-onset disorder that affects the central and peripheral nervous systems and is characterized by cognitive impairment, pyramidal tetraparesis and peripheral neuropathy.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GBE1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge