Anti-GBGT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-13301R-HRP
BOSSBS-13301R-HRPEA 900 EUR InStock
BOSSBS-13301R-HRP
Anti-GBGT1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
GBGT1 is a 347 amino acid single-pass type II membrane protein that belongs to the glycosyltransferase 6 family. Localizing to the golgi apparatus membrane, GBGT1 is widely expressed, with high levels found in placenta, ovary and peripheral blood leukocyte, and lower levels expressed in liver, thymus and testis. GBGT1 utilizes manganese as a cofactor, and assists in the addition of N-acetylgalactosamine (GalNAc) in alpha-1,3-linkage to various substrates, resulting in the formation of glycolipids. Glycolipids are present in most eukaryotic cells and may assist in the adherence of certain pathogens. Existing as two alternatively spliced isoforms, the gene encoding GBGT1 maps to human chromosome 9q34.2 and mouse chromosome 2 A3.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: GBGT1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge