Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GCHFR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13313R-FITC
BOSSBS-13313R-FITCEA 900 EUR InStock
BOSSBS-13313R-FITC
Anti-GCHFR Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
GTP cyclohydrolase I feedback regulatory protein (GFRP) is encoded by the gene GCHFR. GFRP mediates feedback inhibition of GTP cyclohydrolase I activity by tetrahydrobiopterin. GFRP also acts as a mediator for the stimulatory effect of phenylalanine on enzyme activity. L-phenylalanine reverses this inhibition. The molecular weight of GFRP is approximately 9.5kDa although cross-linking experiments have shown that GFRP is usually found as a homodimer or as a pentamer. 15q15.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GCHFR
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge