Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GCM2 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13314R-CY7
BOSSBS-13314R-CY7EA 900 EUR InStock
BOSSBS-13314R-CY7
Anti-GCM2 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Glial cells missing homolog 2 (GCM2), also known as Chorion-specific transcription factor GCMb, is a 506 amino acid nuclear protein. GCM2 is a transcription factor that acts as an essential regulator of parathyroid development. GCM2 is also thought to mediate the effect of calcium on parathyroid hormone expression and secretion in parathyroid cells. GCM2 contains one N-terminal GCM domain, which has DNA binding activity. Mutations of the gene that encodes GCM2 are associated with hypoparathyroidism, an autosomal recessive condition characterized by hypocalcemia and hyperphosphatemia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GCM2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge