Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GCNT7 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13319R-CY7
BOSSBS-13319R-CY7EA 900 EUR InStock
BOSSBS-13319R-CY7
Anti-GCNT7 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Belonging to the glycosyltransferase 14 family, GCNT7, also known as Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 7, is a 430 amino acid glycosyltransferase that is localized to the Golgi apparatus. Other members of this family include GCNT1, GCNT2, GCNT3, GCNT4 and GCNT6. GCNT1 has been shown to play an important regulatory role in the biosynthesis of mucin-type O-glycans, which serve as ligands in cell adhesion. Specifically, GCNT1 expression in leukocytes regulates the synthesis of core 2 O-glycans that carry sialyl-Lewis x (sLex) oligosaccharides, which confer high affinity binding to Selectin proteins. Since downregulation of Selectin ligand expression has been shown to inhibit tissue infiltration, glycosyltransferase 14 family members represent potential drug targets for the treatment of inflammatory disorders and other pathologies involving Selectin proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GCNT7
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge