Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GCSH Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13323R-A750
BOSSBS-13323R-A750EA 890 EUR InStock
BOSSBS-13323R-A750
Anti-GCSH Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
GCSH is a 173 amino acid mitochondrial protein that contains one lipoyl-binding domain and belongs to the gcvH family. Defects in the gene encoding GCSH are the cause of glycine encephalopathy (GCE), an autosomal recessive disease that is also referred to as non-ketotic hyperglycinemia (NKH). Characterized by severe neurological symptoms, patients with GCE have a large amount of glycine accumulated in their body fluids. The gene encoding GCSH maps to human chromosome 16, which encodes over 900 genes and comprises nearly 3% of the human genome.

Type: Primary
Antigen: GCSH
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-173/173
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge