Anti-GDI1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Lieferant: Bioss
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
BS-13324R-A488
BOSSBS-13324R-A488EA 876 EUR InStock
BOSSBS-13324R-A488
Anti-GDI1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Rab proteins, a family of Ras-related small GTP-binding proteins, play a key role in regulating intracellular vesicle trafficking (1?). Rab GDP dissociation inhibitor (Rab GDI or GDI2) forms a soluble complex with Rab proteins and thereby prevents the exchange of GDP for GTP (1?). In mammals, there exist two major isoforms, Rab GDI alpha (also known as XAP-4) and Rab GDI beta (1,4). While the mammalian Rab GDI beta-genes are ubiquitously expressed, the Rab GDI ?genes are predominantly brain-specific (1). Since it is expressed predominantly in neural and sensory tissues, Rab GDI a may serve a specific function in neural signal transmission (5). The gene sequences for the Rab GDI proteins are extremely conserved in evolution, with substantial homology preserved across three eukaryotic kingdoms (5).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GDI1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge