Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GDI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13324R-FITC
BOSSBS-13324R-FITCEA 883 EUR InStock
BOSSBS-13324R-FITC
Anti-GDI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Rab proteins, a family of Ras-related small GTP-binding proteins, play a key role in regulating intracellular vesicle trafficking (1?). Rab GDP dissociation inhibitor (Rab GDI or GDI2) forms a soluble complex with Rab proteins and thereby prevents the exchange of GDP for GTP (1?). In mammals, there exist two major isoforms, Rab GDI alpha (also known as XAP-4) and Rab GDI beta (1,4). While the mammalian Rab GDI beta-genes are ubiquitously expressed, the Rab GDI ?genes are predominantly brain-specific (1). Since it is expressed predominantly in neural and sensory tissues, Rab GDI a may serve a specific function in neural signal transmission (5). The gene sequences for the Rab GDI proteins are extremely conserved in evolution, with substantial homology preserved across three eukaryotic kingdoms (5).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GDI1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge