Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GDPD1/GDE4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13325R-A680
BOSSBS-13325R-A680EA 890 EUR InStock
BOSSBS-13325R-A680
Anti-GDPD1/GDE4 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
GDE4 is a 314 amino acid cytoplasmic and multi-pass membrane protein that belongs to the glycerophosphoryl diester phosphodiesterase family. Expressed in small intestine, placenta, kidney, ovary, thymus, pancreas, spleen, liver and peripheral blood leukocytes, GDE4 contains one GDPD domain and exists as three alternatively spliced isoforms. GDE4 is encoded by a gene that maps to human chromosome 17, which comprises over 2.5% of the human genome and encodes over 1,200 genes. Two key tumor suppressor genes are associated with chromosome 17, namely, p53 and BRCA1.

Type: Primary
Antigen: GDPD1/GDE4
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 221-314/314
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge