Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GEMIN7 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13330R-CY5
BOSSBS-13330R-CY5EA 900 EUR InStock
BOSSBS-13330R-CY5
Anti-GEMIN7 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Gemin7 is a 131 amino acid protein encoded by the mouse gene Gemin7. Gemin7, along with Gemin6, is a significant component of the the large multiprotein human SMN complex. The SMN complex functions as an assembly machine for small nuclear ribonucleoproteins (snRNPs)-the major components of the spliceosome. The survival of motor neurons (SMN) protein, a product of the disease gene of the common neurodegenerative disease, spinal muscular atrophy, is also part of the SMN complex. Although Gemin6 and Gemin7 have no significant sequence similarity with Sm proteins, both adopt canonical Sm folds. Moreover, Gemin6 and Gemin7 exist as a heterodimer, and interact with each other via an interface similar to that which mediates interactions among the Sm proteins. The Gemin6/Gemin7 complex binds to Sm proteins and might help organize Sm proteins for formation of Sm rings on snRNA targets.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Gemin 7
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge