Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MTERF1 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13340R-CY7
BOSSBS-13340R-CY7EA 900 EUR InStock
BOSSBS-13340R-CY7
Anti-MTERF1 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Members of the mTERF family, including MTERF, MTERFD1, MTERFD2 and MTERFD3, are mitochondrial proteins that are believed to be transcription termination factors. MTERF (mitochondrial transcription termination factor 1) is composed of 399 amino acids and contains three leucine zippers that form a three-stranded coiled-coil that binds to DNA. It has been suggested that only the phosphorylated form of MTERF has transcription termination activity. MTERFD1 is also thought to act as a mitochondrial transcription regulator and is expressed as two isoforms produced by alternative splicing. MTERFD3 is believed to be involved in cell cycle regulation and cell growth by modulating mitochondrial transcription. MTERFD3 is expressed in heart, skeletal muscle, pancreas and liver.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MTERF1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge