Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GGA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13343R-A350
BOSSBS-13343R-A350EA 893 EUR InStock
BOSSBS-13343R-A350
Anti-GGA1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The GGA family of proteins (Golgi-localized, ARF-binding proteins) are ubiquitous coat proteins that facilitate the trafficking of soluble proteins from the trans-Golgi network (TGN) to endosomes/lysosomes by means of interactions with TGN-sorting receptors, ARF (ADP-ribosylation factor), and clathrin (1?). Members of the GGA family, GGA1,GGA2 (also known as VEAR) and GGA3, are multidomain proteins that bind mannose 6-phosphate receptors (MPRs) (1,2,4). GGAs have modular structures with an N-terminal VHS (VPS-27, Hrs, and STAM) domain followed by a GAT (GGA and TOM1) domain, a connecting hinge segment, and a C-terminal GAE (?adaptin ear) domain (5). The amino-terminal VHS domains of GGAs form complexes with the cytoplasmic domains of sorting receptors by recognizing acidic-cluster di-leucine (ACLL) sequences (3). GGA1 and GGA2 do not associate with each other, but they do colocalize on perinuclear membranes (2). The cytosolic domain of memapsin 2, but not that of memapsin 1, binds the VHS domains of GGA1 and GGA2 (6). The human GGA1 gene maps to chromosome 22 and encodes a protein that shares 45% sequence identity with GGA2 and GGA3 (1).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GGA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge