Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-MBOAT4 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13355R
BOSSBS-13355REA 752 EUR InStock
BOSSBS-13355R
Anti-MBOAT4 Rabbit Polyclonal Antibody
Antikörper
MBOAT4 (membrane-bound O-acyltransferase domain-containing protein 4, Ghrelin O-acyltransferase) is a 435 amino acid, multi-pass membrane protein that belongs to the membrane-bound acyltransferase family. MBOAT4 functions as an enzyme that attaches an octanoate fatty acid to serine-3 of ghrelin. Ghrelin is a very small, appetite-stimulating hormone secreted by the food-deprived stomach. MBOAT4 can use a variety of fatty acids as substrates including octanoic acid, decanoic acid and tetradecanoic acid. MBOAT4 expression, consistent with its function, is mainly in the stomach and intestines. Due to its primary function, MBOAT4 is a candidate for obesity and appetite suppression studies.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: MBOAT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge