Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLDC Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13370R-A350
BOSSBS-13370R-A350EA 893 EUR InStock
BOSSBS-13370R-A350
Anti-GLDC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
The glycine cleavage system is comprised of AMT (known as Protein T), GCSH (known as Protein H), DLD (known as Protein L) and GLDC (known as Protein P), all of which work together to catalyze the cleavage and degradation of glycine. GLDC (glycine dehydrogenase ), also known as GCE, GCSP (glycine cleavage system P protein) or HYGN1, is a 1,020 amino acid protein that localizes to the mitochondria and belongs to the gcvP family. GLDC binds to glycine and enables the methylamine group from glycine to be transferred to the Protein T. GLDC exists as a homodimer and utilizes pyridoxal phosphate as a cofactor. Mutations in the gene encoding GLDC leads to nonketotic hyperglycinemia (NKH), also known as glycine encephalopathy (GCE), an autosomal recessive disease characterized by accumulation of a large amount of glycine in body fluid and by severe neurological symptoms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GLDC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge