Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLS2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13376R-A750
BOSSBS-13376R-A750EA 890 EUR InStock
BOSSBS-13376R-A750
Anti-GLS2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Plays an important role in the regulation of glutamine catabolism. Promotes mitochondrial respiration and increases ATP generation in cells by catalyzing the synthesis of glutamate and alpha-ketoglutarate. Increases cellular anti-oxidant function via NADH and glutathione production. May play a role in preventing tumor proliferation.

Type: Primary
Antigen: GLS2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 41-140/602
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge