Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-COLGALT2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13379R-CY5
BOSSBS-13379R-CY5EA 900 EUR InStock
BOSSBS-13379R-CY5
Anti-COLGALT2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GLT25D2 is a 626 amino acid protein that localizes to endoplasmic reticulum lumen and belongs to the glycosyltransferase 25 family. Possessing beta-galactosyltransferase activity, GLT25D2 transports beta-galactose to hydroxylysine residues on collagen but lacks glucosyltransferase activity. GLT25D2 is expressed in skeletal muscle and brain, and is encoded by a gene located on human chromosome 1q25.3, a region associated with the hereditary prostate cancer (HPC1) locus. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson’s disease, Gaucher disease, schizophrenia and Usher syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COLGALT2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge