Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLTSCR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13382R-HRP
BOSSBS-13382R-HRPEA 900 EUR InStock
BOSSBS-13382R-HRP
Anti-GLTSCR2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
The tumor suppressor PTEN plays an essential role in regulating signaling pathways involved in cell growth and apoptosis and is inactivated in a wide variety of tumors. Protein interacting with PTEN carboxyl terminus 1 (PICT-1), also designated p60 or Glioma tumor suppressor candidate region gene 2 protein, binds to the C-terminus of PTEN and regulates its turnover. Five Ser/Thr residues within the C-terminal segment of PTEN, including Ser-380, are phosphorylated upon binding of PTEN to PICT-1 and may contribute to the stabilization of PTEN. PICT-1 is localized to the nucleus and/or nucleolus and is highly expressed in pancreas and heart, but can also be detected in liver, skeletal muscle, placenta and kidney. PICT-1 also interacts with herpes simplex virus 1 regulatory proteins ICP22 and ICP0. The tumor suppressor GLTSCR2 gene, which encodes PICT-1, is located in a 150-kb minimal common deletion region for human gliomas, especially oligodendrogliomas, and maps to human chromosome 19q13.3.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: GLTSCR2/PICT1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge