Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-G6PC2 Rabbit Polyclonal Antibody

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13386R
BOSSBS-13386REA 725 EUR InStock
BOSSBS-13386R
Anti-G6PC2 Rabbit Polyclonal Antibody
Antikörper
Glucose-6-phosphatase (G6Pase), is a multicomponent enzyme system that hydrolyzes glucose-6-phosphate in the final step of gluconeogenesis and gluconeolysis. G6Pase localizes to the endoplasmic reticulum, and while liver, kidney, and intestine are the only tissues that express the first identified isoform, G6Pase-alpha, a second form, designated G6Pase-∫, contributes to blood glucose homeostasis in a wider range of tissues. Islet-specific G-6-Pase catalytic subunit-related protein (IGRP), a homolog of the catalytic subunit of G6Pase, may play a role in the regulation of islet metabolism and in insulin secretion induced by metabolites. The exact catalytic acivity of IGRP is not defined. Identification of inhibitors of IGRP have potential therapeutic benefits for treatment of type 2 diabetes resulting from insulin secretion defects. Structurally, IGRP has been shown to be a glycoprotein held in the endoplasmic reticulum by nine transmembrane domains, which are then degraded in cells through the proteasome pathway generating MHC class I presented peptides.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Glucose 6 phosphatase 2/IGRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge