Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GOX Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13387R-CY5
BOSSBS-13387R-CY5EA 900 EUR InStock
BOSSBS-13387R-CY5
Anti-GOX Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Glucose Oxidase is a dimeric enzyme that binds to β-D-glucose and aids in its oxidation into D-glucono-1,5-lactone which then hydrolyzes to gluconic acid. Flavin adenine dinucleotide (FAD) is a cofactor to Glucose Oxidase that acts as the initial electron acceptor and is required for this oxidation to occur. Glucose Oxidase is a natural preservative found in honey, where it reduces atmospheric oxygen into hydrogen peroxide which acts as an antibacterial barrier. Glucose Oxidase is also commonly used in biosensors in which it conveys levels of glucose by keeping track of the number of electrons passed through the enzyme. In this application, Glucose Oxidase is connected to an electrode and the resulting charge is measured.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glucose Oxidase
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge