Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-Fx1A Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-1338R-CY7
BOSSBS-1338R-CY7EA 883 EUR InStock
BOSSBS-1338R-CY7
Anti-Fx1A Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Binding of anti-Fx1A to Heymann nephritis antigens (HA) on rat glomerular epithelial cells (GECs) in culture leads to capping and disappearance of antigens from the cell surface. This process may contribute to the formation of glomerular subepithelial immune deposits in vivo. The authors differentially extracted GECs to determine whether HA redistribution is mediated by cytoskeletal components. Observations were made by phase-contrast and immunofluorescence microscopy on primary and passaged GECs in monolayer culture and by spectrofluorimetry on GECs in suspension.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Fx1A
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge