Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GSTK1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13399R-CY5
BOSSBS-13399R-CY5EA 900 EUR InStock
BOSSBS-13399R-CY5
Anti-GSTK1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Members of the glutathione S-transferase (GST) family of proteins function in the detoxification of xenobiotics to protect cells against toxicant-induced damage. There are eight families of GST proteins, namely alpha, zeta, theta, kappa, mu, pi, sigma and omega, each of which are composed of proteins that have a variety of functions throughout the cell. GSTK1 (glutathione S-transferase kappa 1), also known as glutathione S-transferase subunit 13 (GST 13-13) or GSTK1-1, is a 226 amino acid ubiquitously expressed protein belonging to the kappa class of the GST superfamily that functions in cellular detoxification. Localizing to peroxisome, GSTK1 exists as a homodimer that catalyzes the conjugation of glutathione to a number of hydrophobic substrates leading to their removal from the cell.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GSTK1/Glutathione S Transferase kappa 1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge