Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GSTT2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13441R-A555
BOSSBS-13441R-A555EA 893 EUR InStock
BOSSBS-13441R-A555
Anti-GSTT2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Glutathione (GSH) is a tripeptide antioxidant which reduces disulfide bonds between cytoplasmic proteins. The constitutive enzyme glutathione reductase transforms glutathione into its reduced state which ultimately can provide a measure of cellular toxicity. GSTT2 (glutathione S-transferase theta-2), also known as GST class-theta-2, is a 244 amino acid enzyme with sulfatase activity that functions in conjugating reduced glutathione to hydrophobic electrophiles. GSTT2 exists as a homodimer in the cytoplasm and is expressed in low levels in the liver and the lung. GSTT2 belongs to the GST superfamily and contains both a GST C-terminal and a GST N-terminal domain. The gene encoding GSTT2 exists on human chromosome 22.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GSTT2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge