Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GSTZ1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13442R-FITC
BOSSBS-13442R-FITCEA 900 EUR InStock
BOSSBS-13442R-FITC
Anti-GSTZ1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
GSTZ1 is a member of the glutathione S transferase (GSTs) super family, encoding multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Several transcript variants of this gene encode multiple protein isoforms. GSTZ1 shows minimal glutathione-conjugating activity with ethacrynic acid and 7-chloro-4-nitrobenz-2-oxa-1,3-diazole and maleylacetoacetate isomerase activity. It has low glutathione peroxidase activity with T butyl and cumene hydroperoxides and is able to catalyze the glutathione dependent oxygenation of dichloroacetic acid to glyoxylic acid. Highest expression in liver followed by kidney, skeletal muscle and brain. Also expressed in melanocytes, synovium, placenta, breast and fetal liver and heart.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glutathione Transferase zeta 1/MAAI
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge