Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GLYCTK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13448R-HRP
BOSSBS-13448R-HRPEA 900 EUR InStock
BOSSBS-13448R-HRP
Anti-GLYCTK Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
GLYCTK is a 523 amino acid protein that is expressed as seven isoforms which are present throughout the body. Localized to the cytoplasm and the mitochondrion in an isoform-specific manner, GLYCTK functions to catalyze the ATP-dependent conversion of (R)-glycerate to 3-(R)-glycerate, thereby playing an important role in neural and skeletal muscle systems. Defects in the gene encoding GLYCTK are the cause of D-glyceric acidemia, an inborn error of amino acid metabolism that is best described as nonketotic hyperglycinemia and is characterized by the excretion of D-glyceric acid in the urine.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: HBEBP2/GLYCTK
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge