Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GMEB1 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13454R-CY5
BOSSBS-13454R-CY5EA 900 EUR InStock
BOSSBS-13454R-CY5
Anti-GMEB1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GMEB-1 is a 573 amino acid protein that contains one SAND domain and is a member of the KDWK family of combinatorial transcription modulators. Localized to both the cytoplasm and the nucleus, GMEB-1 forms a heterodimer with GMEB-2 (Glucocorticoid modulatory element-binding protein 2) and, once associated with GMEB-2, plays a key role in parvovirus DNA replication. In addition, GMEB-1 functions alone as a trans-acting factor that, by binding to glucocorticoid modulatory elements (GMEs) in TAT (tyrosine aminotransferase) promoters, increases intracellular sensitivity to glucocorticoid concentrations. GMEB-1 also interacts with initiator procaspases and, via this interaction, can inhibit caspase-induced apoptosis. Due to alternative splicing events, GMEB-1 is expressed as two isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GMEB1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge