Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GMPPB Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13461R-CY5
BOSSBS-13461R-CY5EA 900 EUR InStock
BOSSBS-13461R-CY5
Anti-GMPPB Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GMPPB is a 360 amino acid protein that belongs to the transferase hexapeptide repeat family and is involved in protein modification pathways. Functioning as a GDP-mannose pyrophosphorylase, GMPPB enzymatically catalyzes the conversion of mannose-1-phosphate and GTP to GDP-mannose and a free phosphate, a reaction that is involved in the production of N-linked oligosaccharides. Defects in the gene encoding GMPPB that cause errors in the glycosylation pathway may lead to congenital disorders of glycosylation (CDG). CDGs are multisystemic diseases that often involve both the central and peripheral nervous systems and are often characterized by endocrine and coagulation disorders. GMPPB is expressed as two isoforms due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GMPPB
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge