Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GNL2 Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13471R-CY3
BOSSBS-13471R-CY3EA 900 EUR InStock
BOSSBS-13471R-CY3
Anti-GNL2 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
GNL2 is a nucleolar guanasine-triphosphate binding protein that is ubiquitously expressed at low levels in almost all tissues. GNL2 is involved in the crucial process of trafficking proteins out of the nucleus. Specifically, it is a GTPase that interacts with the 60s preribosomal subunit in the nucleus and facilitates export of the subunit into the cytoplasm. GTPases are responsible for the hydrolysis of GTP by way of a protein region dubbed the G domain. GTPases are often involved in the translocating proteins through membranes gleaning energy for the activity by hydrolizing GTP. GNL2 shares G domain homology and some functionality with nucleostemin (GNL3), another nuclear GTPase. Highest expression of GNL2 is found in testis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNL2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge