Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GNPDA2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13474R-FITC
BOSSBS-13474R-FITCEA 900 EUR InStock
BOSSBS-13474R-FITC
Anti-GNPDA2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
During fertilization in mammals, the sperm activates the egg by causing an increase in the level of free cytoplasmic calcium concentration. This increased calcium concentration induces a characteristic series of oscillations that trigger egg activation and early embryo development. A hamster protein named oscillin is thought to be involved in this pathway. The enzyme glucosamine-6-phosphate isomerase (GNPI) or deaminase (GNPDA1) and the related protein GNPDA2 are the human homologs of hamster oscillin. GNPDA1 and GNPDA2 catalyze the conversion of GNP to fructose-6-phosphate and ammonia. Both proteins exist as homohexamers and are ubiquitously expressed with highest expression in testis, ovary and heart. Three isoforms of GNPDA2 are expressed due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNPDA2
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge