Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GNRH2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13478R-A750
BOSSBS-13478R-A750EA 890 EUR InStock
BOSSBS-13478R-A750
Anti-GNRH2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Human reproduction is controlled by the hypothalaic-pituitary gonadal axis laid down early in fetal development. Gonadotropin releasing hormone (GnRH), also known as GnRH-associated peptide, luteinizing hormone releasing hormone (LHRH), luliberin or gonadorelin, is a decapeptide that is an important molecule in the hypothalamic-pituitary-gonadal axis control circuit. GnRH is produced by hypothalamic neurons and secreted in a pulsatile manner into the capillary plexus of the medianeminence. GnRH affects the release of lutenizing hormone and follicle stimulating hormone from gonadotropic cells in the anterior pituitary. In addition to hypothalamic GnRH (GnRH I), a second GnRH form (GnRH II) functions primarily in the midbrain. GnRH is expressed in the acrosomal region of human sperm and in the anterior pituitary tissue and cancer cells. Unlike GnRH I, GnRH II is highly expressed outside the brain, particularly in the kidney, bone marrow and prostate, suggesting that it may have multiple functions. GnRH binds to a specific G protein-coupled receptor in the pituitary to regulate synthesis and secretion of gonadotropins.

Type: Primary
Antigen: GNRH2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-80/120
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge