Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GNS Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13479R-CY5
BOSSBS-13479R-CY5EA 900 EUR InStock
BOSSBS-13479R-CY5
Anti-GNS Rabbit Polyclonal Antibody (Cy5®)
Antikörper
GNS is a 552 amino acid lysosomal enzyme that hydrolyzes the 6-sulfate groups of the N-acetyl-D-glucosamine 6-sulfate units of keratan sulfate and heparan sulfate. A member of the sulfatase family, GNS assists in the catabolism of heparin, and binds calcium as a cofactor. GNS deficiency results in an autosomal recessive lysosomal storage disorder known as mucopolysaccharidosis type IIID (Sanfilippo D syndrome), which is characterized by mild somatic disease and severe degeneration of the central nervous system. Subject to post-translational internal peptidase cleavage, GNS is encoded by a gene mapping to human chromosome 12q14.2 and mouse chromosome 10 D2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GNS/Glucosamine 6 sulfatase
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge