Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GOLGA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13480R-A488
BOSSBS-13480R-A488EA 893 EUR InStock
BOSSBS-13480R-A488
Anti-GOLGA3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
The Golgi apparatus consists of a series of stacked, flattened membrane stacks called cisternae that are involved in the transport of lipids and proteins in the secretory pathway and are important for Golgi-microtubule interaction. Golgin 160 is a 1,498 amino acid protein that localizes to both the cytoplasm and to the Golgi apparatus and contains a series of coiled-coil domains. Expressed in a variety of tissues, including heart, liver, testis, kidney, lung and salivary gland, golgin 160 functions as a homodimer that interacts with GOLGA7 and is thought to be involved in maintaining Golgi structure and may play a role in nuclear transport and Golgi apparatus localization. Multiple isoforms of golgin 160 exist due to alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Golgin 160
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge