Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GOLGA4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13487R-HRP
BOSSBS-13487R-HRPEA 900 EUR InStock
BOSSBS-13487R-HRP
Anti-GOLGA4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Golgin 245 is also known as p230, GCP2, GOLG or golgi autoantigen and is a 2,230 amino acid protein that is expressed as three isoforms. Golgin 245 is localized to the cytoplasm in cells and is a member of the golgin family, all of which are peripheral membrane proteins associated with the Golgi complex. Golgin 245 has a carboxyl-terminal GRIP domain, which attaches to the trans-Golgi network (TGN) and TGN-derived vesicles. It is thought that the interaction that takes place between golgin 245 and MACF1, which cross-links microtubules to the Actin cytoskeleton, allows proteins to be transported from the TGN to the cell periphery. against golgin 245 are associated with two chronic disorders known as Sjé°ƒren's syndrome and Hepatitis B.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: Golgin subfamily A member 4/Golgin 245
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge