Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-GORASP2 Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:

BS-13492R-CY5
BOSSBS-13492R-CY5EA 900 EUR InStock
BOSSBS-13492R-CY5
Anti-GORASP2 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The Golgi apparatus is a highly complex organelle comprised of a stack of cisternal membranes on the secretory pathway from the ER to the cell surface. The structure is maintained by an exoskeleton or Golgi matrix constructed from a family of coiled-coil protein, the golgins and other peripheral membrane components such as GRASP55 and GRASP65 (1). GRASP55 (Golgi reassembly stacking protien or p59) is a component of the Golgi stacking machinery. GRASP55 is highly homologous to GRASP65 and contains two PDZ domains. GRASP55 is myristoylated and palmitoylated. Unlike GRASP65, GRASP55 does not have detectable binding with the vesicle docking protein GM130 and is located on the medial-Golgi rather than cis-Golgi. Both GRASP55 and GRASP65 function in the stacking of Golgi Cisternae (2,3). The novel coiled-coil protein golgin 45 interacts with GRASP55 and the GTP form of Rab 2, suggesting that GRASP55 and golgin 45 form a Rab 2 effector complex on medial-Golgi essential for normal protein transport and Golgi structure (4). ERK2 directly phosphorylates GRASP55, which is phosphorylated in mitotic cells, suggesting that mitogen-activated protein kinase kinase (MKK)/ERK pathway phosphorylates the Golgi during mitosis (5).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Golgi phosphoprotein 6
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge